- Antimicrobial prophylaxis for infants at risk for recurrent acute otitis media.
Antimicrobial prophylaxis for infants at risk for recurrent acute otitis media.
Vaccine (2001-02-13)
D W Teele, J O Klein, B M Word, B A Rosner, S Starobin, R Earle, C S Ertel, G Fisch, R Michaels, R Heppen, N P Strause
PMID11163478
MATERIALIEN