Merck
  • Role of K+ channels in N-acetylprocainamide-induced relaxation of bovine tracheal smooth muscle.
Role of K+ channels in N-acetylprocainamide-induced relaxation of bovine tracheal smooth muscle.
European journal of pharmacology (2001-03-14)
T Nakahara, H Moriuchi, Y Tanaka, M Yunoki, Y Kubota, K Sakamato, K Shigenobu, K Ishii
ZUSAMMENFASSUNG
We examined the relaxant effects of N-acetylprocainamide, the major hepatic metabolite of procainamide, on bovine tracheal smooth muscle, focusing on the possible involvement of K+ channels. N-acetylprocainamide produced a concentration-dependent and full inhibition of the tension development elicited by methacholine (0.3 or 1 microM). The potency of N-acetylprocainamide in diminishing methacholine-elicited tension development was one-half of that of procainamide. By comparison, N-acetylprocainamide inhibited high-K+ (40 mM)-induced contraction more potently than procainamide though both inhibitions were largely reduced when compared to those against methacholine-induced contraction. Iberiotoxin (30 nM), Ba(2+) (1 mM) or a combination of both agents significantly attenuated the relaxant effect of N-acetylprocainamide on methacholine-induced contraction, whereas apamin (100 nM), 4-aminopyridine (300 microM), and glibenclamide (10 microM) did not affect it. These results suggest that N-acetylprocainamide, similar to procainamide, elicits tracheal smooth muscle relaxation mainly through the activation of plasma membrane K+ channels.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Acetylprocainamid, ≥99%
Sigma-Aldrich
N-Acetylprocainamid -hydrochlorid, ≥99% (HPLC), powder