Merck
  • Optimum timing of insecticide applications against diamondback moth Plutella xylostella in cole crops using threshold catches in sex pheromone traps.
Optimum timing of insecticide applications against diamondback moth Plutella xylostella in cole crops using threshold catches in sex pheromone traps.
Pest management science (2001-07-18)
G V Reddy, A Guerrero
ZUSAMMENFASSUNG
Field trials were conducted in cabbage (Brassica oleracea var capitata), cauliflower (B oleracea var botrytis) and knol khol (B oleracea gongylodes) crops at two different locations in Karnataka State (India) to optimize the timing of insecticide applications to control the diamondback moth, Plutella xylostella, using sex pheromone traps. Our results indicate that applications of cartap hydrochloride as insecticide during a 12-24 h period after the pheromone traps had caught on average 8, 12 and 16 males per trap per night in cabbage, cauliflower and knol khol, respectively, were significantly more effective than regular insecticide sprays at 7, 9, 12 or 15 days after transplantation. This was demonstrated by estimation of the mean number of eggs and larvae per plant, the percentage of holes produced, as well as the marketable yield of the three crops at each location. A good correlation between the immature stages, infestation level, the estimated crop yield and the number of moths caught in pheromone traps was also found, indicating the usefulness of pheromone-based monitoring traps to predict population densities of the pest.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Cartap -hydrochlorid, PESTANAL®, analytical standard