Merck
Antioxidant principles from Bauhinia tarapotensis.
Journal of natural products (2001-07-28)
A Braca, N De Tommasi, L Di Bari, C Pizza, M Politi, I Morelli
ZUSAMMENFASSUNG
A new cyclohexenone (1) and a new caffeoyl ester derivative (2), together with the known compounds (-)-isolariciresinol 3-alpha-O-beta-D-glucopyranoside (3), (+)-1-hydroxypinoresinol 1-O-beta-D-glucopyranoside (4), isoacteoside (5), luteolin 4'-O-beta-D-glucopyranoside (6), and indole-3-carboxylic acid (7), were isolated from the leaves of Bauhinia tarapotensis. The structures of these new compounds were determined by spectroscopic data analysis. The antioxidant activities of 1-7 were determined by measuring their free radical scavenging effects, using the 1,1-diphenyl-2-dipicrylhydrazyl free radical (DPPH) and Trolox equivalent antioxidant activity (TEAC) methods, and the coupled oxidation of beta-carotene and linoleic acid. Compounds 3-5 showed good activities in the DPPH and TEAC tests, while compounds 1 and 2 were active in the coupled oxidation of beta-carotene and linoleic acid bioassay.