Merck
HCN and chemical evolution: the possible role of cyano compounds in prebiotic synthesis.
Tetrahedron (1984-01-01)
J P Ferris, W J Hagan
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Diaminomaleonitril, 98%