Merck
  • Investigation of the herbicide glyphosate and the plant growth regulators chlormequat and mepiquat in cereals produced in Denmark.
Investigation of the herbicide glyphosate and the plant growth regulators chlormequat and mepiquat in cereals produced in Denmark.
Food additives and contaminants (2001-09-25)
K Granby, M Vahl
ZUSAMMENFASSUNG
An LC-MS/MS method for analysing glyphosate and aminomethylphosphonic acid (AMPA) in cereals was developed. The method is based on extraction with water and detection of the ions from the fragmentation m/z 170-->88 (glyphosate) and m/z 112-->30 (AMPA), using electrospray interface in the positive mode. Investigation from the harvests of 1998 and 1999 showed residues of glyphosate and/or its degradation product AMPA in more than half of the cereal samples produced in Denmark. The average concentration of glyphosate in 46 samples from the 1999 harvest was 0.11 mg/kg compared with 0.08 mg/kg for the 1998 harvest (n = 49). Thus, the figures were well below the maximum residue limit (MRL) and no violations were observed. The plant growth regulators chlormequat and/or mepiquat were investigated in cereals from the Danish harvest of 1999 where 83% of the samples contained chlormequat (n = 46) compared with 87% of the samples from the 1997 harvest (n = 52). The average concentration of chlormequat in 1999 was 0.32 mg/kg compared with 0.23 mg/kg in 1997. At 2.9 mg/kg, one sample of wheat bran was exceeding the MRL of 2 mg/kg for wheat. The intakes of the pesticides through the diet of cereals were estimated to comprise 0.04% of the acceptable daily intake (ADI) for glyphosate and 1% of the ADI for chlormequat for an adult Dane.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Mepiquatchlorid, PESTANAL®, analytical standard