Merck
[Studies on the voltammetric behaviour of barbaloin and its application].
Yao xue xue bao = Acta pharmaceutica Sinica (2001-10-13)
Q L Li, L Ren, J B Hu, R Q Feng
ZUSAMMENFASSUNG
In 0.1 mol.L-1 H2SO4 solution, a well-defined reduction wave of barbaloin was obtained by single-sweep osciflopolarography. The peak potential was shown to be -0.75 V (vs SCE). The peak current is proportional to the concentration of barbaloin over the range of 2.0 x 10(-7)-6.0 x 10(-6) mol.L-1. The detection limit is 2.0 x 10(-7) mol.L-1. The voltammetric behaviour of the system was studied by linear sweep and cyclic voltammetry. The adsorption coefficient beta was 1.91 x 10(5). The interaction factor alpha was 1.12. The system is a reversible adsorption wave.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aloin, from Aloe barbadensis Miller leaves, ≥97%
Sigma-Aldrich
Aloin, from Curacao aloe, ~50%