Merck
Circumvention of acquired resistance to doxorubicin in K562 human leukemia cells by oxatomide.
Biological & pharmaceutical bulletin (2001-10-20)
M Ishikawa, R Fujita, S Furusawa, M Takayanagi, K Sasaki, S Satoh
ZUSAMMENFASSUNG
We studied the effect of oxatomide, an antiallergic drug, on the resistance of K562 cells to doxorubicin. Oxatomide synergistically potentiated the cytotoxicity of doxorubicin in doxorubicin-resistant K562 cells (K562/DXR) at a concentration of 1-10 microM, but had hardly any synergistic effect on the parental cell line (K562) at the same concentration. Oxatomide inhibit P-glycoprotein pump-efflux activity and the binding of [3H]-azidopine to the cell-surface protein P-glycoprotein, in a dose-related manner. These results indicate that oxatomide reverses the multidrug-resistance phenotype through direct interaction with P-glycoprotein.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oxatomide, ≥99%