Merck
  • Determination of phenol and resorcinol using 1,3-dibromo-5,5-dimethylhydantoin (DBH) analogous to the Koppeschaar reaction. Analytical methods of pharmacopoeias with DBH in respect to environmental and economical concern, Part 6.
Determination of phenol and resorcinol using 1,3-dibromo-5,5-dimethylhydantoin (DBH) analogous to the Koppeschaar reaction. Analytical methods of pharmacopoeias with DBH in respect to environmental and economical concern, Part 6.
Die Pharmazie (2001-10-31)
M Hilp
ZUSAMMENFASSUNG
PH. EUR. 1997 determines phenol and resorcinol according to the Koppeschaar reaction using hepatotoxic and environmentally hazardous chloroform. Chlorinated hydrocarbons are not required when applying 1,3-dibromo-5,5-dimethylhydantoin (DBH). The reaction time for phenol can be reduced from 45 to 5 min. Blind values according to PH. EUR. 1997 result a systematic error of about -1.8%.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Dibrom-5,5-dimethylhydantoin, 98%