Merck
Contact allergy to IPPD and diphenylthiourea in an orthopaedic brace.
Contact dermatitis (2001-11-28)
C G Aplin, C Bower, K Finucane, J E Sansom
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N′-Diphenylthioharnstoff, 98%