Merck
Electrochemical reduction of chloridazon at mercury electrodes, and its analytical application.
Fresenius' journal of analytical chemistry (2002-01-05)
M Zimpl, M Kotoucek, K Lemr, J Veselá, J Skopalová
ZUSAMMENFASSUNG
The electrochemical behavior of the herbicide chloridazon, I (pyrazon), at different pH is described. The electrode reaction (one wave in acidic media and another in alkaline media), investigated using direct current and pulse voltammetry, controlled-potential coulometry, and HPLC-MS, is a combination of the electroreduction (two-electron in the first step) and a kinetic process as a result of which simple compounds (HCl, NH3) are released and, moreover, a five-membered pyrrole cycle is formed in strongly acid media. Products of the kinetic reaction are further reducible. The dissociation constant of I, pKa = 2.96, was found spectrophotometrically. Fast-scan differential pulse voltammetry (FSDPV) was used for determination of I; the detection limit was 2.7 x 10(-8) mol L(-1) (0.006 microg L(-1)) at pH 2.3. Chloridazon was determined in spiked drinking and river water.