Merck
Methemoglobin formation in human erythrocytes by nitroaromatic explosives.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2002-02-12)
A Maroziene, R Kliukiene, J Sarlauskas, N Cenas
ZUSAMMENFASSUNG
We have examined the structure-activity relationships in methemoglobin (MetHb) formation by high explosives 2,4,6-trinitrotoluene (TNT), 2,4,6-trinitrophenyl-N-nitramine (tetryl) and 2,4,6-trinitrophenyl-N-nitraminoethylnitrate (pentryl), and a number of model nitrobenzenes. In lysed human erythrocytes the rate constants of oxyhemoglobin (OxyHb) oxidation increased with an increase in single-electron reduction potential (E(1)7) or with a decrease of the enthalpies of single-electron reduction of nitroaromatics. Tetryl and pentryl oxidized OxyHb almost 3 times faster than TNT. Although the initial rates of MetHb formation in intact erythrocytes by tetryl, pentryl, and TNT matched their order of reactivity in the oxidation of OxyHb in lysed erythrocytes, TNT was a more efficient MetHb forming agent than tetryl and pentryl during a 24-h incubation. The decreased efficiency of tetryl and pentryl was attributed to their reaction with intraerythrocyte reduced glutathione (GSH) producing 2,4,6-trinitrophenyl-Sglutathione, which acted as a less efficient OxyHb oxidizing agent.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,2-Dinitrobenzol, 97%