- Determination of iodine values according to Hanus using 1,3-dibromo-5,5-dimethylhydantoin (DBH): analytical methods of pharmacopeias with DBH: part 7.
Determination of iodine values according to Hanus using 1,3-dibromo-5,5-dimethylhydantoin (DBH): analytical methods of pharmacopeias with DBH: part 7.
Journal of pharmaceutical and biomedical analysis (2002-02-28)
Manfred Hilp
PMID11861111
ZUSAMMENFASSUNG
USP/NF 2000 [The United States Pharmacopeia, Rockville USA, 24th ed., 2000, p. 1868, The National Formulary, 19th ed. 2000] and PH. EUR. 1997 [European Pharmacopoeia, third ed., Council of Europe, Strasbourg, 1997, pp. 63-64] determine the iodine values according to Hanus with iodine monobromide in glacial acetic acid. This reagent can be replaced by a solution of 1,3-dibromo-5,5-dimethylhydantoin (DBH) and potassium iodide or iodine in the same solvent. Both reagents yield equivalent results by means of method comparison according to Passing and Bablok [J. Clin. Chem. Clin. Biochem. 21 (1983) 709; 22, (1984) 431] in relation to the official method of PH. EUR.
MATERIALIEN