Merck
  • Synthesis of (R)-1,3-butanediol by enantioselective oxidation using whole recombinant Escherichia coli cells expressing (S)-specific secondary alcohol dehydrogenase.
Synthesis of (R)-1,3-butanediol by enantioselective oxidation using whole recombinant Escherichia coli cells expressing (S)-specific secondary alcohol dehydrogenase.
Bioscience, biotechnology, and biochemistry (2002-05-31)
Hiroaki Yamamoto, Akinobu Matsuyama, Yoshinori Kobayashi
ZUSAMMENFASSUNG
The synthesis of (R)-1,3-butanediol (BDO) from its racemate was studied using whole cells of recombinant Escherichia coli expressing an (S)-specific secondary alcohol dehydrogenase (CpSADH) from Candida parapsilosis by enantioselective oxidation. Under the optimized conditions, the yield of (R)-1,3-BDO reached 72.6 g/l, with a molar recovery yield of 48.4% from a racemate of 15% and an optical purity of 95% ee.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(±)-1,3-Butandiol, ReagentPlus®, 99.5%
Sigma-Aldrich
(±)-1,3-Butandiol, anhydrous, ≥99%
Sigma-Aldrich
(R)-(−)-1,3-Butandiol, 95%