Merck
  • Design, synthesis and QSAR studies on N-aryl heteroarylisopropanolamines, a new class of non-peptidic HIV-1 protease inhibitors.
Design, synthesis and QSAR studies on N-aryl heteroarylisopropanolamines, a new class of non-peptidic HIV-1 protease inhibitors.
Bioorganic & medicinal chemistry (2002-06-12)
Roberto Di Santo, Roberta Costi, Marino Artico, Silvio Massa, Rino Ragno, Garland R Marshall, Paolo La Colla
ZUSAMMENFASSUNG
A series of N-aryl heteroarylisopropanolamines in which an indole or a 3-arylpyrrole moiety was linked to an aryl group through an isopropanolamine linker, were designed and synthesized as potential anti-HIV-1-PR agents. Series was tested for their ability in blocking PR activity. As a rule, indole derivatives of class 1 exhibited more potency than pyrrole analogues of class 2 while tert-butylamide substituents increased anti-PR potency. In fact, bis tert-butylamide 1e showed the highest activity with IC(50)=25 microM. Even if not very potent, a simple class of anti-PR agents, with a facile synthetic pathway was discovered. QSAR studies on isopropanolamines 1 and 2 were performed in comparison with diarylbutanols, a new class of non peptidic anti-PR agents, recently discovered by Agouron Pharmaceuticals. QSAR and CoMFA models based on 30 diarylbutanols used as a training set were developed. The obtained models were used to investigate the binding mode of the newly synthesized derivatives 1 and 2. The results of this study suggest that N-aryl heteroarylisopropanolamines bind to the PR active site similarly to the diarylbutanols of Agouron.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Amino-2-propanol, 93%