Merck
  • Effect of the branched-chain alpha-ketoacids accumulating in maple syrup urine disease on the high molecular weight neurofilament subunit (NF-H) in rat cerebral cortex.
Effect of the branched-chain alpha-ketoacids accumulating in maple syrup urine disease on the high molecular weight neurofilament subunit (NF-H) in rat cerebral cortex.
Metabolic brain disease (2002-06-27)
R Pessoa-Pureur, C Funchal, P de Lima Pelaez, L Vivian, S Oliveira Loureiro, R de Freitas Miranda, M Wajner
ZUSAMMENFASSUNG
In this study we investigated the effects of the branched chain alpha-ketoacids accumulating in maple syrup urine disease (MSUD) on the concentrations of the high molecular weight neurofilament subunit (NF-H) associated with the cytoskeletal fraction of the cerebral cortex of 12-day-old rats. Cortical slices were incubated with alpha-ketoisocaproic acid (KIC), alpha-keto beta-methylvaleric acid (KMV) and alpha-ketoisovaleric acid (KIV) at concentrations ranging from 0.5 to 1.0 mM. The cytoskeletal fraction was extracted and the immunoreactivity for phosphorylated and total NF-H was analyzed by immunoblotting. The in vitro 32P incorporation into NF-H was also determined. Results showed that treatment of tissue slices induced with KMV increased Triton-insoluble phosphorylated NF-H immunoreactivity, with no alteration in total NF-H immunoreactivity. Furthermore, KIC treatment drastically increased the total amount of NF-H, whereas KIV did not change either phosphorylated or total NF-H immunoreactivity. KMV also increased the in vitro 32P incorporation into NF-H, confirming the highly phosphorylated NF-H levels detected in the immunoblot. These findings demonstrate that KIC and KMV alter the dynamic regulation of NF-H assembly in the cytoskeletal fraction. Therefore we may suggest that cytoskeletal disorganization may be one of the factors associated with the neurodegeneration characteristic of MSUD disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(±)-3-Methyl-2-Oxovaleriansäure Natriumsalz
Sigma-Aldrich
3-Methyl-2-oxopentansäure, ≥98%