Merck
  • Determination of the acid dissociation constant of bromocresol green and cresol red in water/AOT/isooctane reverse micelles by multiple linear regression and extended principal component analysis.
Determination of the acid dissociation constant of bromocresol green and cresol red in water/AOT/isooctane reverse micelles by multiple linear regression and extended principal component analysis.
Annali di chimica (2002-07-20)
Maurizio Caselli, Annarosa Mangone, Paola Paolillo, Angela Traini
ZUSAMMENFASSUNG
The pKa of 3',3",5',5"tetrabromo-m-cresolsulfonephtalein (Bromocresol Green) and o-cresolsulphonephtalein (Cresol Red) was spectrophotometrically measured in a water/AOT/isooctane microemulsion in the presence of a series of buffers carrying different charges at different water/surfactant ratios. Extended Principal Component Analysis was used for a precise determination of the apparent pKa and of the spectra of the acid and base forms of the dye. The apparent pKa of dyes in water-in-oil microemulsions depends on the charge of the acid and base forms of the buffers present in the water pool. Combination with multiple linear regression increases the precision. Results are discussed taking into account the profile of the electrostatic potential in the water pool and the possible partition of the indicator between the aqueous core and the surfactant. The pKa corrected for these effects are independent of w0 and are close to the value of the pKa in bulk water. On the basis of a tentative hypothesis it is possible to calculate the true pKa of the buffer in the pool.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kresolrot, indicator grade, Dye content 95 %
Sigma-Aldrich
Kresolrot Natriumsalz, indicator grade