Merck
  • Electromagnetic re-warming of cryopreserved tissues: effect of choice of cryoprotectant and sample shape on uniformity of heating.
Electromagnetic re-warming of cryopreserved tissues: effect of choice of cryoprotectant and sample shape on uniformity of heating.
Physics in medicine and biology (2002-08-08)
Martin P Robinson, Monica C Wusteman, Lihong Wang, David E Pegg
ZUSAMMENFASSUNG
A method that has been proposed for the cryopreservation of tissues and organs is to add a cryoprotective agent (CPA) in sufficient concentration to allow vitrification, and to use rapid electromagnetic heating to prevent the formation of ice crystals during the re-warming. We have compared the physical and biological properties of four CPAs, measuring the speed and uniformity of heating in a 36 mm sphere placed in a 434 MHz applicator, and the toxicity to ECV304 endothelial cells. Ethanediol and dimethyl sulfoxide were found to be suitable for rapid, uniform heating but toxic to the endothelial cells at vitrifying concentrations. Butane-2,3-diol was less toxic, but the heating patterns were unacceptably non-uniform. Propane-1,2-diol was not significantly more toxic than butane-2,3-diol, and did allow uniform heating. It is therefore the best choice of CPA for the vitrification of tissues. We have shown that the uniformity of heating correlates with the dielectric properties of the perfusate. Furthermore, we have shown that uniform heating is feasible in non-spherical samples provided they are approximately ellipsoidal.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,2-Ethandithiol, ≥98.0% (GC)
Sigma-Aldrich
1,2-Ethandithiol, technical grade, ≥90%