Merck
  • Adult and paediatric contact immunotherapy with squaric acid dibutylester (SADBE) for recurrent, multiple, resistant, mucocutaneous anogenital warts.
Adult and paediatric contact immunotherapy with squaric acid dibutylester (SADBE) for recurrent, multiple, resistant, mucocutaneous anogenital warts.
Sexually transmitted infections (2002-08-16)
F Dall' Oglio, M R Nasca, O D'Agata, G Micali
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,4-Dibutoxy-3-cyclobuten-1,2-dion, 98%