Merck
[Contents of barbaloin-related compounds in aloe drinks and their change during storage].
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2002-09-20)
Tetsuya Shindo, Hirofumi Ushiyama, Kimiko Kan, Shin-ichi Uehara, Kazuo Yasuda, Ichiro Takano, Kazuo Saito
ZUSAMMENFASSUNG
The contents of barbaloin (BA), isoBA, aloin-dimers A, B, C, D and aloe-emodin (AE) in aloe drinks were investigated. BA and isoBA were detected in 30 of the 31 samples at the levels of 120-570 micrograms/mL and 120-580 micrograms/mL, respectively. Aloin-dimers A, B, C and D were detected in 8 of the 31 samples at the levels of 12-38 micrograms/mL, 13-39 micrograms/mL, 11-36 micrograms/mL and 16-69 micrograms/mL, respectively. AE was detected in all of the 31 samples at the levels of 0.03-1.3 micrograms/mL. When aloe drinks were stored for 4 weeks at 5 degrees C after opening the bottle, decrease of BA and isoBA, and increase of AE and aloin-dimers A, B, C and D were observed in most cases. However, in a few aloe drinks, all of BA, isoBA, aloin-dimers A, B, C, D and AE decreased. In these drinks, the existence of aloin-trimer was elucidated by LC/MS analyses. These data suggested that BA in aloe drinks is converted to the dimer and then to the trimer during storage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aloin, from Aloe barbadensis Miller leaves, ≥97%
Sigma-Aldrich
Aloin, from Curacao aloe, ~50%