Merck
  • Field response of Ips paraconfusus, Dendroctonus brevicomis, and their predators to 2-methyl-3-buten-2-ol, a novel alcohol emitted by ponderosa pine.
Field response of Ips paraconfusus, Dendroctonus brevicomis, and their predators to 2-methyl-3-buten-2-ol, a novel alcohol emitted by ponderosa pine.
Journal of chemical ecology (2002-10-10)
Dennis W Gray
ZUSAMMENFASSUNG
Methylbutenol (MBO) is a major component of the aggregation pheromone of the European spruce beetle Ips typographus and also has been found to be emitted in large amounts by several species of pine native to western North America. This study investigates the influence this signal may have on the behavior of North American bark beetles and examines whether MBO functions as a defensive compound for emitting pines. The response of two North American bark beetles (Ips paraconfusus and Dendroctonus brevicomis) and their predaceous beetles (Trogositidae and Cleridae) to MBO, pheromone, and monoterpenes in varying release rates was investigated in the field using Lindgren funnel traps. MBO exhibited no repellent properties when tested alone, nor did MBO appear to have any effect on the aggregation response of these bark beetles and their predators to their pheromones. These results provide no support for a defensive function of MBO.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Methyl-3-buten-2-ol, 98%