- Highly enantioselective Ag(i)-catalyzed [3 + 2] cycloaddition of azomethine ylides.
Highly enantioselective Ag(i)-catalyzed [3 + 2] cycloaddition of azomethine ylides.
Journal of the American Chemical Society (2002-11-07)
James M Longmire, Bin Wang, Xumu Zhang
PMID12418889
ZUSAMMENFASSUNG
A highly reactive Ag(I)-catalyzed [3 + 2] cycloaddition of azomethine ylides is founded using AgOAc as the catalytic precursor and phosphines as ligands. Using a new bis-ferrocenyl amide phosphine (FAP) as the ligand, we found that high enantioselectivities (up to 97% ee) have been achieved in the [3 + 2] cycloaddition of azomethine ylides. Up to four stereogenic centers can be established in this multicomponent coupling reaction from readily available materials such as aldehydes, aminoesters, and dipolarophiles.
MATERIALIEN