Merck
  • [Effect of simulated microgravity conditions on auxin polar transport of transgenic Arabidopsis].
[Effect of simulated microgravity conditions on auxin polar transport of transgenic Arabidopsis].
Uchu Seibutsu Kagaku (2003-02-04)
T Shimazu, K Miyamoto, J Ueda
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Indol-3-acetamid, 98%