- [Analysis of chemical constituents in the essential oil from Evodia fargesii by GC/MS].
[Analysis of chemical constituents in the essential oil from Evodia fargesii by GC/MS].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2003-02-11)
J Zhang, G Feng, Y Luo
PMID12575060
ZUSAMMENFASSUNG
In the essential oil from Evodia fargesii Dode, 33 constituents were identified by means of GC-MS-DS. The content of these compounds are 91.0% in the essential oil of this plant. The main compounds are eighteen terpenoids.
MATERIALIEN