Merck
Production of D-(--)-3-hydroxyalkanoic acid by recombinant Escherichia coli.
FEMS microbiology letters (2003-02-14)
Kai Zhao, Ge Tian, Zhong Zheng, Jin-Chun Chen, Guo-Qiang Chen
ZUSAMMENFASSUNG
Pathways for extracellular production of chiral D-(-)-3-hydroxybutyric acid (3HB) and D-(-)-3-hydroxyalkanoic acid (mcl-3HA) were constructed by co-expression of genes of beta-ketothiolase (phbA), acetoacetyl-CoA reductase (phbB) and 3-hydroxyacyl-ACP CoA transacylase (phaG), respectively, in Escherichia coli strain DH5alpha. The effect of acrylic acid and glucose on production of both 3HB and mcl-3HA was investigated. It was found that the addition of acrylic acid significantly increased production of 3HB and mcl-3HA consisting of 3-hydroxyoctanoic acid and 3-hydroxydecanoic acid in a ratio of 1:3 from 199 mg x l(-1) to 661 mg x l(-1) and from 27 mg x l(-1) to 135 mg x l(-1), respectively, in shake flask studies when glucose was present in the medium at the very beginning of fermentation. The timing of glucose addition had no effect on 3HB production. In contrast, mcl-3HA production was affected by glucose addition, an mcl-3HA concentration of 193 mg x l(-1) was obtained when glucose was added to the culture at 12 h. A more than seven-fold increase was obtained when compared with that in medium containing glucose at the beginning of fermentation. However, a decrease in production of 3HB and mcl-3HA was found when glucose was added at 12 h to the culture containing acrylic acid. The repressive effect of acrylic acid on acetic acid production was also evaluated and discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(±)-3-Hydroxydecanoic acid, ≥98%