Merck
Plant chitinase as a possible biocontrol agent for use instead of chemical fungicides.
Bioscience, biotechnology, and biochemistry (2003-03-07)
Shuji Karasuda, Shuhei Tanaka, Hiroshi Kajihara, Yuji Yamamoto, Daizo Koga
ZUSAMMENFASSUNG
We investigated whether a plant chitinase can be used as a biocontrol agent instead of chemical fungicides by spraying chitinase E (family 19; class IV) from a yam (Dioscorea opposita Thunb) alone or together with beta-1,3-glucanase directly onto the surface of a powdery mildew infecting strawberry berries and leaves. Results were observed by eye and with a scanning electron microscope. The powdery mildew infecting the strawberries was degraded, mainly by the chitinase, and the disease did not appear again for more than 2 weeks. These results indicated that this kind of plant chitinase might be safe and biodegradable biocontrol agent for use instead of conventional fungicides.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lyticase aus Arthrobacter luteus, lyophilized powder, ≥2,000 units/mg protein, Protein ≥20 % by biuret
Sigma-Aldrich
Lyticase aus Arthrobacter luteus, lyophilized powder, ≥200 units/mg solid