Merck
  • Detection of resistance to fungicides, mating types and fitness of Phytophthora infestans in Hebei, China.
Detection of resistance to fungicides, mating types and fitness of Phytophthora infestans in Hebei, China.
Mededelingen (Rijksuniversiteit te Gent. Fakulteit van de Landbouwkundige en Toegepaste Biologische Wetenschappen) (2003-04-19)
Wen-Qiao Wang, Zhi-Qiang Ma, Xiu-Ying Han, Xiao-Feng Zhang, Shu-Long Chen
ZUSAMMENFASSUNG
In vitro, isolates resistant to metalaxyl (M) and oxadixyl (O) of Phytophthora infestans were 11.2% of 62 isolates from potato and tomato in Hebei Province, mean resistance factor was 15,022 fold and 24,733 fold, respectively, no isolates resistant either to cymoxanil (C) or to dimethomorph (D) were detected. On the other hand, in vivo, isolates resistant to M and O were 29.0% and 32.7%, respectively, among 217 isolates from potato plants in Weichang and Chongli, Hebei, only one isolate resistant to M and O was found among 88 isolates from tomato plants in Xushui, Hebei. Among 73 isolates from potato in Weichang and Chongli, 6 isolates with A2 mating type were 8.2%, 3 A2 isolates resistant to metalaxyl and oxadixyl. 3 A2 isolates were 6.8% of 44 isolates from tomato in Xushui. Cross resistance in Phytophthora infestans was confirmed between M and O, but no cross resistance between C or D and M or O. The field isolates resistant to M and O were outstandingly fitter than sensitive or intermediate ones, mainly due to stronger sporulation capacity of the field isolates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Dimethomorph, PESTANAL®, analytical standard
Supelco
Cymoxanil, PESTANAL®, analytical standard
Supelco
Oxadixyl, PESTANAL®, analytical standard