Merck
  • Characterisation of gene expression patterns in 22RV1 cells for determination of environmental androgenic/antiandrogenic compounds.
Characterisation of gene expression patterns in 22RV1 cells for determination of environmental androgenic/antiandrogenic compounds.
The Journal of steroid biochemistry and molecular biology (2003-04-25)
A Hartel, A Didier, M W Pfaffl, Heinrich H D Meyer
ZUSAMMENFASSUNG
Alteration of androgen receptor function due to hormonally active compounds in the environment, may be responsible for impaired reproductive function in aquatic wildlife. Based on human prostate carcinoma 22RV1 cells, a cell culture expression system was established to test effects of putative androgenic/antiandrogenic compounds on endogenous gene expression. 22RV1 cells were shown to express human androgen receptor, but not human progestin (hPR) or human oestrogen receptor (hER) alpha and beta. Six androgen-regulated genes (ARGs) were chosen to determine androgenic/antiandrogenic action using highly sensitive real-time RT-PCR. Results showed that gene expression is altered in a time-dependent manner. After stimulation of cells by DHT (10nM), synthetic androgen R1881 (1 nM), or organic pesticides (difenoconazole, fentinacetate, tetramethrin) TMPRSS2 mRNA expression was down-regulated by the factor 0.6 after 24h of DHT treatment. Similar results were obtained when cells were assayed for mRNA expression of PSA after fentinacetate and R1881 stimulation. In contrast, TMPRSS2 expression was up-regulated by the factor 0.9 when cells were stimulated by tetramethrin. Final goal of the work is a sensitive determination of differential gene expression by different compounds under study, achievement of substance-specific expression patterns and function related analysis of potential androgens/antiandrogens.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Tetramethrin, PESTANAL®, analytical standard
Supelco
Difenoconazol, PESTANAL®, analytical standard