Merck
  • Interactions between nematophagous fungi and consequences for their potential as biological agents for the control of potato cyst nematodes.
Interactions between nematophagous fungi and consequences for their potential as biological agents for the control of potato cyst nematodes.
Mycological research (2003-05-09)
Helen Jacobs, Simon N Gray, David H Crump
ZUSAMMENFASSUNG
The efficacies of three nematophagous fungi, Paecilomyces lilacinus, Plectosphaerella cucumerina and Pochonia chlamydosporia, for controlling potato cyst nematodes (PCN) as part of an Integrated Pest Management (IPM) regime were studied. The compatibility of the nematophagous fungi with commonly used chemical pesticides and their ability to compete with the soil fungi Rhizoctonia solani, Chaetomium globosum, Fusarium oxysporum, Penicillium bilaii and Trichoderma harzianum were tested in vitro. Paecilomyces lilacinus was the most successful competitor when the ability to grow and inhibit growth of an opposing colony at both 10 and 20 degrees C was considered. P. lilacinus also showed potential for control of the soil-borne fungal pathogen R. solani, releasing a diffusable substance in vitro which inhibited its growth and caused morphological abnormalities in its hyphae. Pochonia chlamydosporia was least susceptible to growth inhibition by other fungi at 20 degrees in vitro, but the isolate tested did not grow at 10 degrees. Plectosphaerella cucumerina was a poor saprophytic competitor. Radial growth of Paecilomyces lilacinus and Plectosphaerella cucumerina was slowed, but not prevented, when grown on potato dextrose agar incorporating the fungicides fenpiclonil and tolclofos-methyl, and was not inhibited by the addition of pencycuron or the nematicide oxamyl. Radial growth of Pochonia chlamydosporia was partially inhibited by all the chemical pesticides tested. The efficacy of Paecilomyces lilacinus as a control agent for R. solani was further investigated in situ. Treatment with P. lilacinus significantly reduced the symptoms of Rhizoctonia disease on potato stems in a pot trial. The effectiveness of P. lilacinus and P. cucumerina against PCN was also tested in situ. Three application methods were compared; incorporating the fungi into alginate pellets, Terra-Green inoculated with the fungi and applying conidia directly to the tubers. Both formulations containing P. lilacinus and formulation mixtures alone, particularly alginate pellets, significantly reduced multiplication of PCN in soil. We conclude that P. lilacinus showed the greatest potential for use in combination with selected fungicides and nematicides as part of an IPM programme for the control of PCN, but further work is required to confirm whether it is effective against PCN in soil.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Oxamyl, PESTANAL®, analytical standard
Supelco
Pencycuron, PESTANAL®, analytical standard