Merck
Control of synaptic strength, a novel function of Akt.
Neuron (2003-06-24)
Qinghua Wang, Lidong Liu, Lin Pei, William Ju, Gholamreza Ahmadian, Jie Lu, Yushan Wang, Fang Liu, Yu Tian Wang
ZUSAMMENFASSUNG
Akt (also known as PKB), a serine/threonine kinase involved in diverse signal-transduction pathways, is highly expressed in the brain. Akt is known to have a strong antiapoptotic action and thereby to be critically involved in neuronal survival, but its potential role in the dynamic modulation of synaptic transmission is unknown. Here we report that Akt phosphorylates, both in vitro and in vivo, the type A gamma-aminobutyric acid receptor (GABA(A)R), the principal receptor mediating fast inhibitory synaptic transmission in the mammalian brain. Akt-mediated phosphorylation increases the number of GABA(A)Rs on the plasma membrane surface, thereby increasing the receptor-mediated synaptic transmission in neurons. These results identify the GABA(A)R as a novel substrate of Akt, thereby linking Akt to the regulation of synaptic strength. This work also provides evidence for the rapid regulation of neurotransmitter receptor numbers in the postsynaptic domain by direct receptor phosphorylation as an important means of producing synaptic plasticity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphor(V)-oxychlorid, ReagentPlus®, 99%
Sigma-Aldrich
Phosphor(V)-oxychlorid, 99.999%