Merck
  • Comparison of different fluorimetric HPLC methods for analysis of acidic polyether toxins in marine phytoplankton.
Comparison of different fluorimetric HPLC methods for analysis of acidic polyether toxins in marine phytoplankton.
Analytical and bioanalytical chemistry (2003-10-11)
M J Nogueiras, Ana Gago-Martínez, Antonio I Paniello, Marian Twohig, Kevin J James, James F Lawrence
ZUSAMMENFASSUNG
The human toxic syndrome, diarrhetic shellfish poisoning (DSP), is caused by polyether toxins that are present in bivalve molluscs but originate from some species of marine phytoplankton. During the last few years different HPLC methods with fluorescence detection (FLD) have been proposed for analysis of marine toxins, including polyether toxins, in shellfish and phytoplankton. Several derivatization reagents have been proposed in the literature, with the aim of converting the acidic DSP toxins into their corresponding fluorescent derivatives. In this work we report results obtained from HPLC-FLD analysis of extracts from phytoplankton, including Dinophysis spp.,harvested off the south-west coast of Ireland. Three different reagents were used for fluorescent derivatization: 3-bromomethyl-6,7-dimethoxy-1-methyl-2(1H)-quinoxalinone (BrDMEQ), 9-chloromethylanthracene (CA), and "in situ" 9-anthracenyldiazomethane (ADAM). Derivatization was performed under conditions previously optimised. The DSP derivatives were cleaned using different SPE procedures then analysed by HPLC-FLD. In this study, the use of BrDMEQ, CA, and "in situ" ADAM was compared in terms of sensitivity and selectivity. Evaluation of HPLC methods for analysis of DSP toxin derivatives was also conducted; the presence of okadaic acid (OA), dinophysistoxin-2 (DTX-2), and pectenotoxin-2 seco acids (PTX1SAs) was detected in the sample extracts studied.