Merck
  • Survey of chloropropanols in soy sauces and related products purchased in the UK in 2000 and 2002.
Survey of chloropropanols in soy sauces and related products purchased in the UK in 2000 and 2002.
Food additives and contaminants (2003-11-05)
C Crews, S Hasnip, S Chapman, P Hough, N Potter, J Todd, P Brereton, W Matthews
ZUSAMMENFASSUNG
The results of surveys to investigate the levels of 3-monochloropropane-1,2-diol (3-MCPD) and 1,3-dichloropropanol (1,3-DCP) in UK retail samples of soy sauces and similar products are reported. The products, sampled in 2000 and 2002, were analysed for 3-MCPD using an established solvent extraction technique with a reporting limit of 0.01 mg kg(-1), which also detected 2-monochloropropane-1,2-diol (2-MCPD), and for 1,3-DCP by an automated headspace method with a reporting limit of 0.005 mg kg(-1), which also detected 2,3-dichloropropanol (2,3-DCP). In the 2000 survey, 3-MCPD was quantified in 32 of 100 samples. After normalization to 40% dry matter, it was quantified at or above 0.02 mg kg(-1) in 25 of the samples and in excess of 1 mg kg(-1) in 16 samples, the highest containing 82.8 mg kg(-1). 2-MCPD was found in 26 samples, at up to 17.6 mg kg(-1) after normalization to 40% dry matter. The presence of 1,3-DCP was detected in 17 of the samples, at levels between 0.006 and 0.345 mg kg(-1). 1,3-DCP was only detected where 3-MCPD was present, but the levels of 1,3-DCP and 3-MCPD were not correlated. 2,3-DCP was detected in 11 samples at levels ranging from 0.006 to 0.043 mg kg(-1). In the 2002 survey, 3-MCPD was quantified (> 0.01 mg kg(-1)) in only eight of 99 samples and 2-MCPD in three samples. After normalization to 40% dry matter, 3-MCPD was present at or above 0.02 mg kg(-1) in seven of these, the maximum level being 35.9 mg kg(-1). 1,3-DCP was detected in this sample alone, at 0.017 mg kg(-1).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Dichlor-2-propanol, 97%