Merck
Study of charge transfer complexes of menadione (vitamin K3) with a series of anilines.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2003-12-13)
Purnendu Pal, Avijit Saha, Asok K Mukherjee, Dulal C Mukherjee
ZUSAMMENFASSUNG
Menadione (vitamin K(3)) has been shown to form charge transfer complexes with N,N-dimethyl aniline, N,N-dimethyl p-toluidine and N,N-dimethyl m-toluidine in CCl(4) medium. The CT transition energies are well correlated with the ionisation potentials of the anilines. The formation constants of the complexes have been determined at a number of temperatures from which the enthalpies and entropies of formation have been obtained. The formation constants exhibit a very good linear free energy relationship (Hammett) at all the temperatures studied.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
m-Toluidin, 99%