Merck
[Elucidation of pharmacological mechanism of drugs by studying the receptors].
The Japanese journal of antibiotics (2003-12-19)
Hiroshi Handa
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
E3330, ≥98% (HPLC)