- Solid state study of hydrogen bonding in dichlorophen crystals.
Solid state study of hydrogen bonding in dichlorophen crystals.
International journal of pharmaceutics (2003-12-31)
Philip J Cox, Nicola M Foote, Stephen M MacManus
PMID14698572
ZUSAMMENFASSUNG
An X-ray crystallographic study of dichlorophen has been performed. Intramolecular hydrogen bonding is found within the molecule and intermolecular hydrogen bonding is present between molecules. The formation of dimers within the crystal lattice has been established.
MATERIALIEN