Merck
  • Analysis of mitosis in squash preparations of larval brains: orcein, Giemsa, Hoechst 33258, DAPI, quinacrine, and N-banding.
Analysis of mitosis in squash preparations of larval brains: orcein, Giemsa, Hoechst 33258, DAPI, quinacrine, and N-banding.
Methods in molecular biology (Clifton, N.J.) (2004-01-07)
Laura Fanti, Sergio Pimpinelli
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Orcein, synthetic
Sigma-Aldrich
Orcein, synthetic, certified by the BSC