Merck
[Examination of sex-hormonal activity of some additives for PVDC film].
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2004-01-22)
Minoru Ohta, Shozo Oshima, Toshio Iwasa, Naofumi Ito, Masashi Morii, Masayoshi Morino, Tadashi Nakamura, Kenji Nagai
ZUSAMMENFASSUNG
Stabilizers (epoxidized linseed oil and epoxidized soybean oil) and plasticizers (acetyl tributyl citrate, diacetyl monolauryl glyceride and dibutyl sebacate) commonly used in polyvinylidene chloride (PVDC) films and extracts of such films were investigated for estrogenic and androgenic activity by means of estrogen receptor (ER) and androgen receptor (AR) competitive ligand-binding assays. Further, in in vivo experiments, ovariectomized Sprague-Dawley rats were observed for uterine wet weight change, uterine endometrium hyperplasia and vaginal mucosa cornification, following administration of each test compound or extract orally (0.5 or 500 mg/kg) or subcutaneously (0.5 or 100 mg/kg). No significant response or change was observed with any of the test compounds or extracts, either in vitro or in vivo. The results thus indicate that neither the stabilizers and plasticizers used in PVDC films, nor their extracts, exert sex-hormonal activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tributyl-O-acetylcitrat, 98%
Sigma-Aldrich
Dibutylsebacat, ≥97.0% (GC)
Sigma-Aldrich
Tributyl-2-Acetylcitrat, ≥98%, FG
Sigma-Aldrich
Dibutylsebacat, technical grade
Supelco
Dibutylsebacat, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tributyl-2-Acetylcitrat, Pharmaceutical Secondary Standard; Certified Reference Material