Merck
  • Binding and transport of [3H](2S,4R)- 4-methylglutamate, a new ligand for glutamate transporters, demonstrate labeling of EAAT1 in cultured murine astrocytes.
Binding and transport of [3H](2S,4R)- 4-methylglutamate, a new ligand for glutamate transporters, demonstrate labeling of EAAT1 in cultured murine astrocytes.
Journal of neuroscience research (2004-03-03)
Karina Apricò, Philip M Beart, Duncan Crawford, Ross D O'Shea
ZUSAMMENFASSUNG
Transporters for L-glutamate (excitatory amino acid transporters; EAATs), localized to astrocytes, are involved intimately in intermediary metabolism within the brain. Because (2S,4R)-4-methylglutamate (4MG) has affinity for glial EAATs, we employed [(3)H]4MG to define the characteristics of EAATs in cultured murine astrocytes and describe new approaches to analyze EAAT function. Specific binding of [(3)H]4MG in astrocytic membranes at 4 degrees C represented 90% of total binding. Binding was rapid (apparent t(1/2) approximately 7 min) and saturable. Saturation and Scatchard analyses indicated a single binding site (n(H) = 0.8) with a K(d) of 6.0 +/- 1.5 microM and B(max) = 9.7 +/- 2.9 pmol/mg protein. Binding of [(3)H]4MG to astrocytic homogenates was Na(+)-dependent and inhibited by K(+). Compounds acting at EAATs, such as L-glutamate (Glu), D-aspartate (D-Asp), L-(2S,3S,4R)-2-(carboxycyclopropyl)glycine and L-trans-pyrrolidine-2,4-dicarboxylate displaced binding to nonspecific levels. L-Serine-O-sulphate, an EAAT1-preferring ligand, fully displaced binding of [(3)H]4MG. In contrast, inhibitors having preferential affinity for EAAT2, L-threo-3-methylglutamate, dihydrokainate, and kainate, were relatively ineffective binding displacers. Agonists and antagonists for Glu receptors failed to significantly inhibit [(3)H]4MG binding. Studies with [(3)H]D-Asp reinforced evidence that [(3)H]4MG was binding to EAATs. These data were consistent with Western blot analyses, which indicated abundant expression of EAAT1 but not EAAT2. [(3)H]4MG was also accumulated rapidly (apparent t(1/2) approximately 4 min) into whole astrocytes by a sodium- and temperature-sensitive process (K(m) of 146 +/- 24 microM, V(max) = 336 +/- 27 nmol/mg protein/min), which possessed an EAAT1-like pharmacologic profile. These findings confirm that 4MG is a substrate for EAAT1 and that the binding assay developed using [(3)H]4MG can be utilized in various preparations including cultured astrocytes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(2S,3R,4S)-α-(Carboxycyclopropyl)glycine, solid