Merck
Trace explosives detection by photoluminescence.
TheScientificWorldJournal (2004-03-09)
E Roland Menzel, Kimberly K Bouldin, Russell H Murdock
ZUSAMMENFASSUNG
Some field tests in counter-terrorism efforts to detect explosive traces employ chemistries that yield colored products. We have examined a test kit of this kind, ETK(Plus), based on widely used chemistries and employed extensively by the Israel Police. Our investigation focuses on the prospect of gaining sensitivity by replacing the normal colorimetric modality with photoluminescence detection, which, to our knowledge, has not been explored to date. We find two or more orders of magnitude sensitivity gains for all explosives studied, using field-worthy photoluminescence techniques. We have also investigated a general lanthanide-based photoluminescence approach which shows promise and the ability to photoluminescence-detect trace explosives in the presence of intense background color and/or background fluorescence by time-resolved imaging.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ferroin-Indikator, 0.1 wt. % in H2O