Merck
  • Different binding thermodynamics of Ni2+, Cu2+, and Zn2+ to bacitracin A1 determined by capillary electrophoresis.
Different binding thermodynamics of Ni2+, Cu2+, and Zn2+ to bacitracin A1 determined by capillary electrophoresis.
Electrophoresis (2004-03-09)
Massimo Castagnola, Diana V Rossetti, Rosanna Inzitari, Alessandro Lupi, Cecilia Zuppi, Tiziana Cabras, Maria B Fadda, Giuseppina Onnis, Raffaele Petruzzelli, Bruno Giardina, Irene Messana
ZUSAMMENFASSUNG
Thermodynamics of the binding of Ni(2+), Cu(2+) and Zn(2+) to bacitracin A(1) was studied by capillary electrophoresis measuring the peptide effective mobility at different pH in the presence of increasing concentration of the three ligands. The affinity follows the order Ni(2+) > Cu(2+) > Zn(2+), with association constant values of (2.3 +/- 0.1)x10(4), (4.9 +/- 0.2)x10(3), and (1.5 +/- 0.1)x10(3) M(-1), respectively. The only model able to rationalize mobility data implies that metal ion binds to the P(0) peptide form. Moreover, mobility values indicated a change of bacitracin A(1) acidic properties on Ni(2+) and Cu(2+) binding, with a shift of the pK(a) of N-terminal Ile-1 from 7.6 to about 5 and of the pK(a) of the delta-amino group of D-Orn-7 from 9.7 to about 7. Even though on Zn(2+) binding a shift of the N-terminal Ile-1 pK(a) was observed, restrictions in the pH range suitable for investigation, due to precipitation phenomena, did not allow establish if the shift of D-Orn-7 lateral chain pK(a) also occurred. Nonetheless, if present, the shift should be limited to the 7.8-9.7 range. Mobility data indicated that the Stokes radius of the complexes is ca. 3 A lower than that of the free peptide. The present results indicate that metal-ion binding to bacitracin A(1) is more complex than previously assumed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Bacitracin A, VETRANAL®, analytical standard, main stereoisomer ~50 %