Merck
Hydrogen abstraction ability of different aromatic nitroxides.
Free radical research (2004-04-06)
Elisabetta Damiani, Paola Astolfi, Massimo Benaglia, Angelo Alberti, Lucedio Greci
ZUSAMMENFASSUNG
Indolinonic aromatic nitroxides have been shown to efficiently inhibit free radical mediated oxidation reactions in biological systems. Since all antioxidants also possess pro-oxidant activity, possibly through a hydrogen abstraction process from suitable substrates, the relative hydrogen abstraction abilities of these compounds were evaluated. Different hydrogen donors were reacted with an indolinic and two indolinonic nitroxides and the rates of hydrogen abstraction were determined using UV-Vis spectroscopy. From the data obtained, a structure-activity relationship was found. In addition, the hydrogen abstraction ability of these compounds was found to be much greater than that of the aliphatic nitroxide TEMPO, despite existing reports indicating that these two classes of compounds show similar antioxidant activities in biological systems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Diethylhydroxylamin, ≥98%