- Cloning, expression and partial characterization of a Haemaphysalis longicornis and a Rhipicephalus appendiculatus glutathione S-transferase.
Cloning, expression and partial characterization of a Haemaphysalis longicornis and a Rhipicephalus appendiculatus glutathione S-transferase.
Insect molecular biology (2004-05-26)
I da Silva Vaz Jnr, S Imamura, K Ohashi, M Onuma
PMID15157234
ZUSAMMENFASSUNG
The ticks Haemaphysalis longicornis and Rhipicephalus appendiculatus are important parasites worldwide. The current method for control of cattle ticks involves the use of chemicals. Nevertheless, parasite resistance is an ever increasing global problem. Glutathione S-transferases (GSTs) play a central role in detoxication of xenobiotic and endogenous compounds. Several authors have noted that an increase in GST activity is associated with resistance to insecticides and acaricides. In the present study, we report the cloning and expression of GST cDNAs from H. longicornis and R. appendiculatus. In addition, we determine the effect of three acaricides (ethion, deltamethrin and diazinon) on the enzymatic activity of rGSTs.
MATERIALIEN