- Versatile asymmetric synthesis of the kavalactones: first synthesis of (+)-kavain.
Versatile asymmetric synthesis of the kavalactones: first synthesis of (+)-kavain.
Organic letters (2004-07-02)
Thomas E Smith, Mabel Djang, Alan J Velander, C Wade Downey, Kathleen A Carroll, Sophie Van Alphen
PMID15228268
ZUSAMMENFASSUNG
[reaction: see text] Three asymmetric pathways to the kavalactones have been developed. The first method is chiral auxiliary-based and utilizes aldol reactions of N-acetyl thiazolidinethiones followed by a malonate displacement/decarboxylation reaction. The second approach uses the asymmetric catalytic Mukaiyama additions of dienolate nucleophile equivalents developed by Carreira and Sato. Finally, tin-substituted intermediates, prepared by either of these routes, can serve as advanced general precursors of kavalactone derivatives via Pd(0)-catalyzed Stille couplings with aryl halides.