- Transverse dephasing optimised NMR spectroscopy in solids: natural-abundance 13C correlation spectra.
Transverse dephasing optimised NMR spectroscopy in solids: natural-abundance 13C correlation spectra.
Chemphyschem : a European journal of chemical physics and physical chemistry (2004-07-16)
Gaël De Paëpe, Anne Lesage, Stefan Steuernagel, Lyndon Emsley
PMID15253313
MATERIALIEN