Merck
Lipophilicity study of salicylamide.
Acta pharmaceutica (Zagreb, Croatia) (2004-07-28)
Marica Medić-Sarić, Ana Mornar, Ivona Jasprica
ZUSAMMENFASSUNG
Molecular lipophilicity was studied using salicylamide as a model drug. Log P value for the target compound was experimentally determined by the shake-flask method and calculated using nine different computer programs based on atom/fragment contributions, structural parameters, atom-type electrotopological-state indices and neural network modeling, or topological structure descriptors. Our analysis demonstrates good agreement between the experimentally observed log P value of salicylamide and the value calculated by the CSLogP program, based on topological structure descriptors and electrotopological indices.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salicylamid, 99%
Sigma-Aldrich
Salicylamid, puriss., ≥99.0% (T)