Merck
  • Antitumor activity of pyrvinium pamoate, 6-(dimethylamino)-2-[2-(2,5-dimethyl-1-phenyl-1H-pyrrol-3-yl)ethenyl]-1-methyl-quinolinium pamoate salt, showing preferential cytotoxicity during glucose starvation.
Antitumor activity of pyrvinium pamoate, 6-(dimethylamino)-2-[2-(2,5-dimethyl-1-phenyl-1H-pyrrol-3-yl)ethenyl]-1-methyl-quinolinium pamoate salt, showing preferential cytotoxicity during glucose starvation.
Cancer science (2004-08-10)
Hiroyasu Esumi, Jie Lu, Yukiko Kurashima, Tomoyuki Hanaoka
ZUSAMMENFASSUNG
An anthelminthic, pyrvinium pamoate (PP), 6-(dimethylamino)-2-[2-(2,5-dimethyl-1-phenyl-1H-pyrrol-3-yl)ethenyl]-1-methyl-quinolinium pamoate salt, has been found to be extremely toxic to PANC-1 cells in glucose-free medium, but not to be toxic to the same cells cultured in ordinary medium, Dulbecco's modified Eagle's medium (DMEM). It showed the same preferential toxicity for various cancer cell lines during glucose starvation. When 0.1 microg/ml PP was added to the medium, spheroid growth of human colon cancer cell line WiDr was strongly inhibited to a diameter of 750 microm, and this finding is consistent with the concept of anti-austerity. PP was also found to exert antitumor activity against human pancreatic cancer cell line PANC-1 in nude mice and SCID mice when it was administered subcutaneously or orally. Regarding the mechanism of PP action, inhibition of Akt phosphorylation, which has been found to be essential for the austerity mechanism, was observed in vitro and in vivo. These findings indicate that PP may be useful for anticancer therapy and that anti-austerity therapy could be a novel strategy for anticancer therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)