Merck
  • Sensitive silver staining of protein in sodium dodecyl sulfate-polyacrylamide gels using an azo dye, calconcarboxylic acid, as a silver-ion sensitizer.
Sensitive silver staining of protein in sodium dodecyl sulfate-polyacrylamide gels using an azo dye, calconcarboxylic acid, as a silver-ion sensitizer.
Electrophoresis (2004-08-10)
Li-Tai Jin, Sun-Young Hwang, Gyurng-Soo Yoo, Jung-Kap Choi
ZUSAMMENFASSUNG
A highly sensitive silver staining method for detecting proteins in sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) was developed. It is based on the silver nitrate staining method but also employs an azo dye, calconcarboxylic acid (NN), as a silver-ion sensitizer. It increases silver binding on protein bands or spots by the formation of a silver-dye complex and also increases the reducing power of silver ions to metallic silver by NN itself with formaldehyde. After a 2 h gel fixing step, the protocol including sensitization, silver-ion impregnation, and reduction steps can be completed in 1 h. The sensitivity is superior to that of silver stain with glutardialdehyde as a silver-ion sensitizer. The detection limit of NN-silver stain is 0.05-0.2 ng protein. Considering the high sensitivity without using glutardialdehyde, the NN-silver stain would be useful for routine silver staining of proteins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Calconcarbonsäure, suitable for use as an indicator for the titration of calcium