Merck
  • Zero-crossing second derivative spectrophotometry for simultaneous determination of palladium and nickel using 1-(2-pyridylazo)-2-naphthol in mixed micellar solutions.
Zero-crossing second derivative spectrophotometry for simultaneous determination of palladium and nickel using 1-(2-pyridylazo)-2-naphthol in mixed micellar solutions.
Annali di chimica (2004-09-07)
Habibollah Eskandari
ZUSAMMENFASSUNG
1-(2-Pyridylazo)-2-naphthol (PAN) has been used for the simultaneous determination of nickel and palladium at trace levels. PAN complexes of nickel and palladium in the pH 1.98 form red and green colored complexes, respectively, which are soluble in aqueous 4:1 Triton X-100 to sodium dodecylsulfate (SDS) micellar media with total detergent concentration of 3.2%. Under optimum conditions, calibration graphs for the simultaneous determination by second derivative spectrophotometry were obtained. Zero crossing second derivative spectrophotometry at 668 and 572 nm, respectively for palladium and nickel was used for the simultaneous determination. The method is able to determine palladium to nickel ratio 70:1 to 1:6 (Wt/Wt), accurately. Accuracy and reproducibility of the determination method on the known various amounts of palladium and nickel in their binary mixtures were tested. Effects of diverse ions on the determination of palladium and nickel to investigate selectivity of the method also were studied.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-(2-Pyridylazo)-2-naphthol, indicator grade