Merck
Adsorption of fluorescein by protein crystals.
Biotechnology and bioengineering (2004-09-08)
A Cvetkovic, M Zomerdijk, A J J Straathof, R Krishna, L A M van der Wielen
ZUSAMMENFASSUNG
Adsorption characteristics of native and cross-linked lysozyme crystals were examined using fluorescein as model adsorbate. The adsorption isotherms exhibited Langmuir or linear behavior. The affinity constant (b1) and the adsorption capacity (Qsat) for fluorescein were found to depend on the type and concentration of co-solute present in the solution. The dynamics of adsorption isotherm transition from Langmuir to linear showed that affinity of lysozyme for solutes increases in the order 2-(cyclohexylamino)ethanesulphonic acid (CHES), 4-morpholinepropanesulphonic acid (MOPS), acetate, fluorescein. Furthermore, the crystal morphology, the degree of cross-linking of the crystals, and, in particular, solution pH were identified as factors determining fluorescein adsorption by the lysozyme crystals. These factors seem to affect crystal capacity for the solute more than affinity for the solute. Adsorption of fluorescein by cross-linked tetragonal lysozyme crystals was exponentially dependent on the lysozyme net charge calculated from the final solution pH. The 3-5-fold increase in the fluorescein adsorption as a result of cross-linking is presumably due to the increasing hydrophobicity of the lysozyme crystal.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioUltra, Molecular Biology, ≥99.5% (titration)