Merck
Simple coupling reaction between amino acids and weakly nucleophilic heteroaromatic amines.
Journal of combinatorial chemistry (2004-09-14)
Gilles Quéléver, Stéphane Burlet, Cédrik Garino, Nicolas Pietrancosta, Younes Laras, Jean-Louis Kraus
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphor(V)-oxychlorid, ReagentPlus®, 99%
Sigma-Aldrich
Phosphor(V)-oxychlorid, 99.999%